Mini Mailers and Mailing Sleeves, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61520-12 61520-40 61521-43 61521-44 61522-01 61522-02 61522-04 61522-34 61522-35 61522-95 61523-02 61523-03 61523-36 61523-41 61523-42 61523-92 61523-96
102093-482EA 164.45 CAD
102093-482 102093-484 102093-486 102093-488 102093-492 102093-494 102093-496 102093-498 102093-500 102093-502 102093-504 102093-506 102093-508 102093-512 102093-514 102093-516 102093-518 102093-520 102093-522
Mini Mailers and Mailing Sleeves, Electron Microscopy Sciences
Mailers and Shippers
Mini mailers either for temperature sensitive specimens or for non-hazardous laboratory specimens.
Mini Mailers for Temperature Sensitive Shipment:
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents
Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants
Insulating, thick wall polystyrene foam construction
Walls on model 440 are 1” thick; walls on model 312 are 1 ½” thick
Model 312 is in corrugated carton
Standard Mini Mailers:
Maximum breakage protection for non-hazardous laboratory specimens. Ideal for biological, environmental, research and development, public health, industries hygiene, and veterinary use
Accommodate tubes, vials, bottles, petri dishes and cans
Lightweight polystyrene foam construction saves postage
Reusable and recyclable
Standard mailing sleeves available