Lab Mailer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61515-07 61515-09 61515-39 61515-41 61515-56 61515-64 61515-69
102093-434EA 64.57 CAD
102093-434 102093-436 102093-438 102093-440 102093-442 102093-444 102093-446 102093-448
Lab Mailer, Electron Microscopy Sciences
Mailers and Shippers
Maximum breakage protection for known non-hazadous blood, serum, tissue, and other specimens.
  • Accommodates VacuTainer® Microtainer® and other collection / transport tubes
  • Available in multi-tube, slide, and tube / slide combinations
  • Lightweight polystyrene foam construction saves postage
  • Reusable and recyclable
VacuTainer® and Microtainer® are trademarks of Becton Dickenson.
ThermoSafe® is a trademark of Tegrant Corporation, ThermoSafe Brands.