Limonene-Mount, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
17987-01
17987-25
17987-06
100496-554EA
440.77
CAD
100496-554
100496-556
100501-606
Limonene-Mount, Electron Microscopy Science
Stains
Microscopy and staining reagents
This mounting medium is made with limonene, a natural product from orange peels. It preserves tissues and cell smears that can be dehydrated with organic solvents in immunohistochemistry, e.g. DAB and DAB with nickel or cobalt.
- A direct substitute for BioMeda™ Clarion Mount
- Use for mounting tissues and cell smears
- Works well with alkaline phosphatase chromogens, and organic solvent resistant Supper Fast Red
- A good choice for mounting H & E stained slides.
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)