Richter Anti-Vibration Pad, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
67118-10 67118-20 67118-30
102095-982EA 75.6 CAD
102095-982 102095-984 102095-986
Richter Anti-Vibration Pad, Electron Microscopy Sciences
Surface Protectors Bench Protectors
Richter Anti-Vibration Pad inexpensively dampens vibrations from lab equipment and creates a stable surface that will keep equipment in place.
  • Fits a variety of bench-top vortexes, shakers, rockers and microscopes
  • Small, lightweight design makes transportation easy
  • Pads can be stacked for heavier units or laid side-by-side for larger equipment