96-Well and 384-Well Color Orienters, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70490-15 70490-16
102096-940EA 104.41 CAD
102096-940 102096-942
96-Well and 384-Well Color Orienters, Electron Microscopy Sciences
Well Plate Stands and Well Orienters
Fits most standard flat and round bottom 96-well and 384-well plates.
  • Bright colors make it easy to distinguish between columns and rows
  • Ideal for use with The Pipetter Guide for efficient and stress-free identification of rows and columns