Disposable Mini Scalpel, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72041-11 72041-14 72041-15
102097-792EA 259.82 CAD
102097-792 102097-794 102097-796
Disposable Mini Scalpel, Electron Microscopy Sciences
Scalpels and Scalpel Assemblies Scalpels
Feather Disposable Mini Scalpels are made from heat-treated stainless steel. The blades are smaller than standard blades, come with chemical resistant, slender plastic handle.
Handle is 145 mm (5 3/4") long. Available in three popular styles: #11, #14 and #15, and packaged in a sterile pouch, 20 per box.
ISO 14001 compliant, ISO 9001 certified and ISO13485 certified.