Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72043-10 72043-11 72043-14 72043-15 72043-20 72043-21 72043-22 72043-25
102097-798EA 89.21 CAD
102097-798 102097-800 102097-802 102097-804 102097-806 102097-808 102097-810 102097-812 102097-818
Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
Scalpels and Scalpel Assemblies Scalpels
Made from heat-treated stainless steel, these scalpels are excellent for delicate precise procedures
  • Heat-treated
  • Comfortable hand fit
  • Retractable shield
  • Color-coded blade size identification
They fit the hand comfortably and are designed with safety in mind. They reduce injury and transmitted disease from contaminated blades. They feature a clear protective, retractable shield and have a color coded button on the protective shield which assists with blade size identification.
Packaging: Individually packaged, 10 per box.