Petri Dish Labels, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70630-100 70630-32 70630-50 70630-70 70631-06 70631-08 70631-12 70631-16
102097-066EA 68.07 CAD
102097-066 102097-068 102097-070 102097-072 102097-074 102097-076 102097-078 102097-080
Petri Dish Labels, Electron Microscopy Sciences
Labels
PetriStickers™ are ideally sized stickers for labeling petri dishes.
  • Label petri dishes quickly, neatly and reliably
  • Labels remain firmly intact in temperatures from 4 to 42 °C
Instantly form an imprint petri dish for quick reference then can remove if need.