Petriseal and Containerseal Tapes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70500-B 70500-C 70500-R 70500-W 70500-Y 70501-R 70501-W 70501-Y 70502-W
102097-030EA 101.93 CAD
102097-030 102097-032 102097-034 102097-036 102097-038 102097-040 102097-042 102097-044 102097-048
Petriseal and Containerseal Tapes, Electron Microscopy Sciences
Tapes
Ideal sealing tapes for petri dishes, microplates, lids, caps, and other lab items.
  • Enjoy all the benefits of Petriseal and Containerseal in a new wider format
  • Containerseal XL makes it easier to seal large containers and bottles and to cover wider gaps
Stretches, conforms, and seals the irregular perimeter of microplates or petri dishes. Waterproof and chemical resistant. Tapes and adhesive resist most solvents and caustic agents. Temperature resistant, seal remains flexible in moderate freezer temperatures and withstands intermittent exposure to 100°C.