Well Orienter, Pad and Stand, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70490-10 70490-18 70490-20 70490-30
102096-932EA 79.19 CAD
102096-932 102096-946 102096-948 102096-950 102096-954
Well Orienter, Pad and Stand, Electron Microscopy Sciences
Well Plate Stands and Well Orienters
The well orienter is used for easy pipetting and identification of samples in microplates. With the well plate stand samples are viewed and pipetted at a comfortable 30° without strain.
  • Convenient visual aid for pipetting and identification of samples in microplates
  • Sharp, alpha-numeric characters compatible with 96-, 48- or 24-well microplates
  • Neoprene pad and rubber feet prevents slippage of the plate and stand
  • No more cross-referencing headaches
The well pad is a convenient note pad that corresponds to the layout of a 96-well plate. It is used to keep accurate records of samples in a microplate.