Sealing Films, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70534-10 70534-20 70536-10 70536-20
102097-052EA 246.02 CAD
102097-052 102097-054 102097-058 102097-060
Sealing Films, Electron Microscopy Sciences
Seals Microplate Seals
Use sealing films to eliminate well-to-well contamination, microbial and viral contamination, and sample spillage. Non breathable membranes prevent evaporation.
  • Sterilized by gamma irradiation and free of cytotoxins
  • Two scored end-tabsmake film positioning and removal hassle-free
  • Thermal Stability to withstand a wide temperature range
  • Pierceable sealing films facilitate easy removal of samples with a pipet tip
Breathe-Easy® is an adhesive, gas-permeable, cell culture membrane. It is made of 25.4 µm thick polyurethane. It is UV transparent to 300 nm and can be used in temperature range of -80 to 100 °C.
Breathe-EASIER™ is a rapid gas transmissable sealing membrane for cell growth. Breathe-Easier membranes permit rapid, consistent gas exchange of Oxygen, CO2 and water vapor for uniform cell. It is made of 139.7 µm thick non-woven rayon. It is not UV transparent and can be used in temperature range of -20 to 80 °C.
AluminaSeal™ is non-cytotoxic and sterile and is resistant to DMSO, chemicals, solvents and other common reagents. It can be used for Cold and PCR Storage in a temperature range of -80 to 110 °C. It is made of 68.6 µm aluminum and is not gas permeable or UV transparent.
HotSeal™ is a PCR sealing film for use in the temperature range of -40 to 125 °C. It is made of 58.4 µm polyester and is not gas permeable or UV transparent.