Thermal Transfer Labels, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77592-1 77592-2 77592-3 77592-4 77592-5 77592-65 77592-70 77592-75 77592-80 77592-81 77592-82 77592-83 77592-84 77592-85 77592-86 77592-87 77592-88
102098-644EA 237.77 CAD
102098-644 102098-646 102098-648 102098-650 102098-652 102098-660 102098-662 102098-664 102098-666 102098-668 102098-670 102098-672 102098-674 102098-676 102098-678 102098-680 102098-682
Thermal Transfer Labels, Electron Microscopy Sciences
Labels
Thermal Transfer Cryo-Tags are a reliable and convenient way to identify samples with text or a barcode on microtubes, microplates, bottles and other laboratory containers. Pre-cut, peel-off labels designed for use in thermal transfer printers. Print popular barcode formats for fast and easy sample identification. Works with all thermal transfer printers.
  • Unique solvent-resistant, acrylic-based adhesive provides excellent cold temperature performance
  • Specially coated for use with a resin ribbon that provides the best performance under extreme conditions
  • Resists Degradation from scuffing, chemicals, moisture and wide temperature fluctuations
  • Thermal Transfer Labels for Storage in Temperatures From −196°C to 80°C
  • Print as little as one label at a time, eliminating label waste and saving time and money
  • Print crisp, clear, professional quality labels