Nunc™ 96 Well Optical Bottom Plates, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64815-10 64816-15 64815-05 64815-15 64815-20
100500-508CS 4836.4 CAD
100500-508 100490-668 100500-506 100490-658 102095-740
Nunc™ 96 Well Optical Bottom Plates, Electron Microscopy Sciences
Microplates Cell Culture Plates
The NUNC™ Coverglass Optical Bottom 96 Well Plates have an optically clear polystyrene plate bottom
This clear base permits microscopic viewing and reading of well The plate’s upper structure is offered in black for fluorometric detection. Sterile. External dimensions 128 x 86 mm.