Narrow Mouth Dewars, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62019-20 62019-50 62019-52
100497-530EA 3451.83 CAD
100497-530 100497-534 100501-626
Narrow Mouth Dewars, Electron Microscopy Sciences
Dewars and Cryogenic Vessels
Spherical Dewar Flask:
Lab grade dewars. Made from borosilicate glass and vacuumed to 10-7 torr, without pads between walls, eliminating heat transfer for maximum efficiency. They come with low evaporation stopper and weighted base.
Spherical Dewar Flask:
Metal Casting does not extend the full length of the flask.  Florence flask design, offers the most efficient way of holding liquid nitrogen.  Available with an optional Fiber Glass Caddie for safety, easy carrying, and decanting liquids from the above flasks.