Illuminated Magnilites® Big Eye™, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68204-00 68202-10
100492-738EA 129.99 CAD
100492-738 100492-736
Illuminated Magnilites® Big Eye™, Electron Microscopy Sciences
Magnifiers
"Big Eye" is one of our largest magnilites® with a 7.5", 3 diopter (1.75x) or a 7.5" 4 diopter (2x) optical quality lens. Wrapped in a ESD safe shade, it includes a 250 cm (10") shadowless 34-watt cirline bulb. Available with cool white or our exclusive "Glare-Free" bulb, 25" or 43" Ultraflex® arms, excellent holding ability with superior clamp (cast iron). Comes with 10 years mechanical warranty.
  • ESD safe construction
  • Ergonomic large lens
Voltage: 120 or 230 V
Certifications: UL/CUL, CE available –230 volt