Tungsten Boat, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
73816-B2 73816-B5 73816 73816-B3 73816-B4 73816-B6 73816-B7
100489-544EA 227.77 CAD
100489-544 100489-548 100501-836 100501-912 100489-546 100489-550 100489-552
Tungsten Boat, Electron Microscopy Sciences
Baskets Wire Baskets
Tungsten has the highest melting point and lowest vapor pressure of all metals, and at the temperatures over 1650°C has the highest tensile strength
Tungsten is the most useful material used in vacuum evaporation technique, due to its special properties and cost factor.
It has excellent corrosion resistance and is attacked only slightly by most mineral acids.
Order Now
SPECIFICATIONS
Environmentally Preferable