Compartment Tray with Sliding Lids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64338-07 64338-18
100502-538EA 18.01 CAD
100502-538 100498-664
Compartment Tray with Sliding Lids, Electron Microscopy Sciences
Trays/Dishes/Pots Laboratory Trays/Dishes
Organizing singular or groups of items in individual compartments. Stored items stay separately and are readily visible at all times. Extra-thick wall construction permits containment of heavy objects; protects delicate items.
Order Now
SPECIFICATIONS
Environmentally Preferable