Pinholes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
PH-C1000 PH-C1 PH-C50 PH-100 PH-10 PH-C250 PH-C2 PH-C25 PH-C5
100495-960EA 693.38 CAD
100495-960 100495-950 100495-962 100495-956 100495-946 100495-958 100495-948 100495-952 100495-954
Pinholes, Electron Microscopy Sciences
Graticules, Reticles and Grids
These pinholes are prepared from pure copper foil, 3 mm in diameter, and 25 microns thick.
  • Possess very high roundness and edge retention
  • Blackened on one surface
  • Mounted in black anodized aluminum discs
These grids are used in spatial filtering, controlling the diameter of light beams, creating point light sources and Image analysis.
Mounted in a recessed hole in an anodized holder, these pin holes measure 16 mm in diameter.