Biopsy Foam Pads, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62325-06 62325-04 62325-01
100490-168EA 79.06 CAD
100490-168 100490-166 100490-164
Biopsy Foam Pads, Electron Microscopy Sciences
Biopsy Pads
A cellular polyester blue foam that has uniform air pockets throughout the pad for optimal fluid flow
It prevents either small or fragmented tissue specimens from being lost. To Use: Place small samples or specimen fragments between the two pads and place the specimen in a cassette or capsule.