Nunc™ 384 Well® Optical Bottom Plates – Polymer Base, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64811-20 64810-35 64810-52 64810-54
100500-498CS 1451.26 CAD
100500-498 100500-490 102095-728 102095-730 102095-732
Nunc™ 384 Well® Optical Bottom Plates – Polymer Base, Electron Microscopy Sciences
Microplates Cell Culture Plates
The NUNC™ Coverglass Optical Bottom 96- and 384- Well Plates have an optically clear polystyrene plate bottom
This clear base permits microscopic viewing and reading of well The plate’s upper structure is offered in black (for fluorometric detection).