Zeiss SEM Mounts, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75500 75510 75520
100488-718EA 99.84 CAD
100488-718 100488-720 100488-722
Zeiss SEM Mounts, Electron Microscopy Sciences
Specimen Mounting Supplies
Flat End Pin Surface
  • Diameter: 12.7 mm (1/2")
  • Slotted Head diameter, 3.1 mm (1/8") Pin