Platinum Crucibles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
66504-11 66504-15 66504-5 66504-8 66505-14 66505-7 66500-3 66501-5 66504-10 66505-11 66505-3 66504-9 66505-8
101413-074EA 6662.1 CAD
101413-074 101413-082 101413-090 101413-096 101413-108 101413-122 101413-062 101413-070 101413-072 101413-102 101413-114 101413-098 101413-124
Platinum Crucibles, Electron Microscopy Sciences
Crucibles
Platinum has excellent resistance to chemical attack by acids and fusion mixtures. This is what has made platinum an essential tool in chemical analysis; flux fusions which are widely used on a routine basis for the preparation of samples for x-ray fluorescence etc. All crucibles are made from platinum with an approximate weight in grams and an approximate in millimeter with a tolerance of ±5% . Lids are sold separately and in single units.
Ordering information: EMS strives to maintain accurate pricing. However, due to fluctuations in precious metal prices, pricing on products containing precious metals is not guaranteed. We will contact you if there is a discrepancy.