Lyophilization Seals, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60320-07 60330-13N 60321-07 60322-13 60322-07
101412-414EA 59.3 CAD
101412-414 101412-432 101412-418 101412-424 101412-422
Lyophilization Seals, Electron Microscopy Sciences
Closures Bottle Closures
The Lyophilization seals come in four styles.
The Center Disc Tear Out style exposes the top of the stopper to the syringe. Natural in color. The Flip-Cap, Center Tear style is easy-to-use. Push upward on plastic button to access stopper or septum. No sharp metal edges. Removal of the Flip Cap Button can be accomplished with one hand, freeing the other for the syringe. The Complete Tear Off style seals can be completely removed and contents poured from the bottle. The Lined Aluminum Seals are ideal for filling or retrieving contents with a syringe and as closures for many instrumentationMore Product Information