Disposable Pipettes – Sterile, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70953-1 70953-5 70954-2 70953-10 70954-10 70953-50
101413-236EA 117.95 CAD
101413-236 101413-244 101413-254 101413-238 101413-250 101413-246 101413-248
Disposable Pipettes – Sterile, Electron Microscopy Sciences
Pipettes Pasteur/Transfer Pipettes
Crystal Grade polystyrene is used for consistency in manufacture and maximum clarity, with easy to read crisp black printed graduations enabling the pipettes to be used accurately to ±1%. Sterile by gamma irradiation. These Pipettes fit all Quick Release Pipette Fillers. These pipettes are tested for pyrogenicity, and have pyrogen levels of less than 0.25 EU/ml.