Fume Hood Shelves, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64396-02 64396-01 64396-03
101413-000EA 435.7 CAD
101413-000 101412-998 101413-002
Fume Hood Shelves, Electron Microscopy Sciences
Furniture Shelves
Special designed fume hood shelves to offer addition storage space for your lab.
  • Designed to maintain air-flow
  • Increase work space under fume hood
  • Available in 3 sizes
Keeps hazardous materials and other items off hood floor for safety.