Adhesive Labels on Rolls, Round, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77580-B 77580-R 77580-Y 77580-G 77580-W 77580-O 77580-44 77580-SI 77581-B 77581-CO 77581-G 77581-GO 77581-LA 77581-MA 77581-O 77581-R 77581-SI 77581-W 77581-Y 77582-100 77582-18 77582-75 77586-20 77586-21 77580-CO 77580-GO 77580-LA 77580-MA 77581-P 77586-10 77586-11
101413-956EA 79.77 CAD
101413-956 101413-962 101413-966 101413-958 101413-964 101413-960 102098-562 102098-572 102098-574 102098-576 102098-578 102098-580 102098-582 102098-584 102098-586 102098-590 102098-592 102098-594 102098-596 102098-598 102098-600 102098-602 102098-610 102098-612 102681-366 102681-368 102681-370 102681-372 102681-374 102681-376 102681-378
Adhesive Labels on Rolls, Round, Electron Microscopy Sciences
Labels
Tough-Spots® pre-cut peel-off round labels are used on microcentrifuge tube tops. The PVC labels are easy to write on and accept writing from most lab markers including solvent resistant pens and teeny writers.
  • In sheets (for laser printer) or on a roll
  • Select from 11 colors
  • A variety of sizes, colors and formats ensures the right label for the job
Ordering information: Rainbow includes blue, green, orange, red, and yellow.
Delivery information: 5 Across Tough-Spots® come in a dispensing box that assists in labeling individual tubes used in small experiments. White or assorted roll with 1000 spots each of blue, green, lavender, red and yellow.