Stirring Rods, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
66200-10 66201-10 66200-15 66200-20 66200-30 66200-40
101413-030EA 29.31 CAD
101413-030 101413-032 102095-948 102095-950 102095-954 102095-958
Stirring Rods, Electron Microscopy Sciences
Stirrers Stirring Rods
These stirring rods are available in two versions: pure 'PTFE' solid rod with tapered end and pure 'PTFE' with bendable steel core.
The version with steel core can be bent into a permanent shape. Rods are inert and will not scratch, use up to 280 °C.