Aluminum Oxide Abrasive Film Discs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50357-05 50357-12 50357-20 50358-3 50358-5 50358-05 50358-1 50358-12 50358-20 50357-03 50357-1 50357-30 50358-9
101411-862EA 175.76 CAD
101411-862 101411-866 101411-868 101411-888 101411-892 101411-880 101411-882 101411-884 101411-886 101411-860 101411-864 101411-872 101411-894
Aluminum Oxide Abrasive Film Discs, Electron Microscopy Sciences
Abrasives
Alumina is widely used in sample grinding and polishing. Our Aluminum Oxide Abrasive film with a PSA back or a plain back are ready to use in all sample polishing applications.