SuperFrost™ Clipped Corner Slide, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71882-10 71882-01
101413-470CS 1242.78 CAD
101413-470 101413-468
SuperFrost™ Clipped Corner Slide, Electron Microscopy Sciences
Slides Microscope Slides
Designed specially for the Sysmex SP-100 slide prep unit and recommended for the Diagnostics.
The 45 degree clipped corner slide offers exceptional cleanliness and resistance to sticking. The tight tolerances for the length and width help to reduce accumulation of glass chips within the instrument. The pre-cleaned slide is 25×75 mm and approximately 1 mm thick. Ideal for technicians, who routinely prepare large number of slides.