Magic Wands, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63323-02 63323-01 63323-03
101412-836EA 34.27 CAD
101412-836 101412-832 101412-834
Magic Wands, Electron Microscopy Sciences
Stirrers Stirring Rods
Magic wands are ideal for use for electronic high-tech areas to laboratories, from preparation of samples to applying adhesives, using it as a stirring rod, or even dosing samples, etc.
  • PEEK polyetheretherketone reinforced with carbon nano
  • Very hard, rigid, high tensile and flexural strength, very high wear resistance
  • High heat capability (260-300°C), good dimension stability, low thermal linear expansion coefficient
  • Excellent resistance to chemicals and aggressive agents, excellent resistance to thermal aging
  • ESD-safe material 106 Ohm
Typical applications include handling of components in cleaning/chemical/assembly processes also at high temperature (soldering).
Probes are 6" (150 mm) long.