Cryo Labels, Laser Printable, on Sheets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77561-05 77560-B 77561-B 77560-05 77560-Y 77561-W 77560-G 77560-W 77560-O 77561-G 77560-R 77561-O 77561-R 77561-Y 77559-W
101413-872EA 192.02 CAD
101413-872 101413-860 101413-874 101413-858 101413-870 101413-882 101413-862 101413-868 101413-864 101413-876 101413-866 101413-878 101413-880 101413-884 102098-498
Cryo Labels, Laser Printable, on Sheets, Electron Microscopy Sciences
Labels
Specially designed innovative labels for cryogenic storage. Made with a flexible, polyolefin label material and acrylic adhesive. This unique combination expands and contracts together with temperature fluctuations resulting in a strong adhesive bond.
  • Withstands freezer temperatures to –196 °C (liquid and vapor phase nitrogen), boiling water baths (100 °C), autoclaving and moderate oven temperatures (150 °C)
  • Resist most organic solvents and caustic agents
  • Adhere to plastics, glass and metals without cracking, peeling or degrading
  • Specially top-coated to accept laser printing and other markings
  • Heat-resistant, stay-flat sheets will not jam in laser printers
Virgin Polypropylene tubes are recommended. Avoid the use of siliconized surfaces.
Packaging: Rainbow packs with 4 sheets of each color: Blue, Green, Red, Yellow.