Cryo Labels, Laser Printable, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77575-C 77576-C
102098-502EA 209.9 CAD
102098-502 102098-504
Cryo Labels, Laser Printable, Electron Microscopy Sciences
Labels
Clear durable cryogenic storage labels allow the contents of vials beneath labels to remain visible.
  • Temperature-resistant –196 to 70 °C
  • Remains fully adhered down to –196 °C (liquid and vapor phase nitrogen)
  • Resist most organic solvents and caustic agents
  • Adhere to plastics, glass and metals without cracking, peeling or degrading
  • pecially top-coated to accept laser printing and other markings
  • Heat-resistant, stay-flat sheets will not jam in laser printers
Made with a flexible, vinyl label material and permanent acrylic adhesive. This unique combination expands and contracts together as temperatures fluctuate resulting in a strong and sustained adhesive bond.