Stirring Hot Plates Accessories, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72351-12 72351-18 72351-21 72351-22 72351-25 72351-30
100493-724EA 158.17 CAD
100493-724 100493-726 100493-728 101413-534 101413-536 101413-538
Stirring Hot Plates Accessories, Electron Microscopy Sciences
Stirrers Stirring Tools
External temperature controller for digital hot plates: Controls the temperature of liquids up to 200 °C in vessels on top of the Pyroceram heated top of Pyroceram heated top
Stainless Steel Support Rod is a stainless steel, two piece, 18" (45.7 cm) support rod with a 5/16 (0.8 cm) diameter that consists of two 9" rods that screw together. It screws into the base of Corning® hot plate and stirrer. Accessory Kit is designed for all Corning digital display hot plates and stirring hot plates. It is voltage independent and includes a temperature controller, a two piece, 5/16" (0.8 cm) diameter, 18" (45.7 cm) long support rod, and magnetic stir bar retriever.
Increases accuracy and repeatability by controlling liquid temperature not top plate temperature. When in use, the controller icon on the display well be lit and the temperature control on the unit can set from 5 to 200 °C. Temperature Probe Support Clamp has two openings 13" (9 mm) and 12" (3.75 mm) for holding probes. It is designed to be used with a boss heat clamp or standard right angle clamp.