4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70075-O 70075-P 70075-PE 70075-T 70075-W 70075-G 70075-L 70075-FP
101413-218EA 714.54 CAD
101413-218 101413-220 101413-222 101413-224 101413-206 101413-226 101413-212 101413-216 101413-208
4-Compartment Micromesh Cassettes, Electron Microscopy Sciences
Embedding Cassettes Biopsy Processing Cassettes
Biopsy processing cassettes with four square compartments each measuring 13 mm.
  • Cover and base have about 1676 square openings maximizing fluid exchange and ensuring proper drainage
Each case contains four dispenser boxes of 250 cassettes with covers.
Order Now
SPECIFICATIONS
Environmentally Preferable