Manomark Pen All Surface Marker, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72127-B1 72127-AS 72127-B12
101413-506EA 25.31 CAD
101413-506 101413-504 102097-872
Manomark Pen All Surface Marker, Electron Microscopy Sciences
Pens Ink Pens
Writes on all plastic, glass, metal, wool, paper and cloth, including "PTFE", fluoropolymer resins
  • With its precision seal valve the pens life is preserved and as well it allows for a smooth flow of ink even if it’s left uncapped
  • Once dry, the solvent base ink is temperature resistant from 0 to 155 °C (32 to 310 °F)
Markings are insoluble in water or hydrocarbons, resist many solvents, and are easily removed with alcohol.