Microsurgical Knife, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72047-30 72047-45 72047-15
101413-488EA 178.19 CAD
101413-488 101413-490 101413-486
Microsurgical Knife, Electron Microscopy Sciences
Scalpels and Scalpel Assemblies Scalpels
Made from tool steel wire. Sharpened to microsurgical quality.
The knife-edges are available in three different configurations: 45, 30 and 15 degrees angle. The handle is plastic and they come sterile and in a blister pack individually.
Packaging: 6 blister packs to a box.