EZFlip Conical Centrifuge Tubes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63030-25 63031-25
101412-662EA 110.95 CAD
101412-662 101412-666
EZFlip Conical Centrifuge Tubes, Electron Microscopy Sciences
Tubes Centrifuge Tubes
EzFlip™ Conical Centrifuge tubes offer a safer alternative to screw cap tubes. A patented closure system improves ergonomics and versatility.
  • Leak-proof - hinged, flip-cap design
  • Tubes are graduated and feature a writing area
  • One handed opening and closing
  • Good chemical resistance
  • Freezer or refrigerator capability
  • Clarified polypropylene
  • Compatible with all major centrifuges and rotors
An improved tube and cap design increases mechanical strength to withstand aggressive centrifugal forces. The 15 ml tube's maximum relative centrifugal force (RCF) is 8500×g, and the 50 ml tube's maximum RCF is 9500×g.
Packaging: All EZFlip™ tubes are available in rack or bag of 50, case of 500.