C-Flat™ Holey Carbon Grids for Cryo TEM, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CF212-25 CF212-50 CF213-100 CF213-25 CF213-50 CF-222C-100 CF-222C-50 CF-224C-100 CF-224C-50 CF412-100 CF412-25 CF412-50 CF413-100 CF413-25 CF413-50 CF422-50 CF442-100 CF442-25 CFT413-50 CFT-224C-50 CFT413-100 CFT-224C-100 CF313-100-AU
102100-052EA 855.03 CAD
102100-052 102100-056 102100-058 102100-060 102100-062 102100-066 102100-070 102100-072 102100-076 102100-094 102100-128 102100-130 102100-134 102100-136 102100-138 102100-140 102100-148 102100-162 102100-164 103306-282 103306-294 103306-850 103306-862 103306-946
C-Flat™ Holey Carbon Grids for Cryo TEM, Electron Microscopy Sciences
Graticules, Reticles and Grids
C-flat™ is an ultra-flat, holey carbon-coated TEM support grid for transmission electron microscopy (TEM).
  • Manufactured without plastics so it is clean and without residue
  • Better particle dispersion and more uniform thickness
  • Use for single particle analysis, cryo electron tomography and automated TEM analysis
C-flat™ leads to better data sets. Made with patent pending technology, C-flat™ provides an ultra-flat surface that results in better particle dispersion and more uniform ice thickness. Patterning is done using deep-UV projection lithography,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable