Anti-Static Foam Tipped Applicator, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72318 72316
100501-826EA 72.96 CAD
100501-826 100489-880
Anti-Static Foam Tipped Applicator, Electron Microscopy Sciences
Swabs Swabs for Cleaning
Choose from swabs that feature an antistatic foam head – or swabs with completely static-free construction.
Static-dissipative swabs attract dirt and dust for easy cleaning, while reducing and dissipating static electricity. Antistatic properties will not decay over time. Swabs meet all industry standards and requirements, including ASTM-D257 and Federal test method 4046, standard #101C.