Cloth-Textured Hi/Lo Temperature Tape, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77012-10 77012-05 77012-07
100488-162EA 146.51 CAD
100488-162 100488-158 100488-160
Cloth-Textured Hi/Lo Temperature Tape, Electron Microscopy Sciences
Tapes
These tapes mold tightly to any container, withstands acids, alkalis, oils and greases.
It's will not crack, peel, or fall off in 550 °F dry oven temperatures. High or low temperature tape also sticks tightly in. Tape is available in widths of ½, ¾, and 1 on 250" roll. This tape must be cut with scissors.