Cross Lines Graticules, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
67983-10 67983-01 67983-20 67983-25 67983-30 67984-16 67984-21 67983-05 67984-19 67983-20.4 67983-23 67983-24.5 67983-025 67983-26 67983-27
100499-012EA 307.52 CAD
100499-012 100499-008 100499-014 100499-016 100499-018 100499-020 100499-024 100499-010 100499-022 103303-852 103303-854 103303-856 103303-858 103303-860 103303-862
Cross Lines Graticules, Electron Microscopy Sciences
Graticules, Reticles and Grids
The microscope graticle is used for measurements in two directions for sighting and alignment.
  • Cross lines with surface chrome image
  • Nominal line width of 0.020, 0.040 or 0.005 mm
  • Covers the entire field of view