CryoVial®, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61800-02 61800-5A 61800-3A 61802-1A 61802-2A 61800-2A 61802-02 61800-4A 61801-5A 61802-3A 61801-02 61801-4A
100500-280CS 755.29 CAD
100500-280 100500-290 100500-286 100500-306 100500-308 100500-284 100500-304 100500-288 100500-294 100500-302 100500-310 100500-292 100502-590
CryoVial®, Electron Microscopy Sciences
Vials
These vials are designed for storage of biological material, human or animal cells, at temperatures as low as –190 °C.
  • The cap features a long skirt for one hand aseptic technique
  • A super fast thread design that allows tightening or removal with a mere 1¼" turn, and an inside thread design that will not contribute to possible contamination
  • Tubes are provided with white marking area for sample identification and can be color-coded by the use of a CAPINSERT®
  • Compatible with most storage systems
  • Sterilized by gamma irradiation
The closures and tubes are both manufactured of polypropylene having the same coefficient of expansion, which further enhances the leak-proof qualities of these vials at changing temperatures
Tubes are available in three series.
Ordering information: All Cryovials® are sold by case quantity (10× 100 per case).
Packaging: Packaged in unique tempered-proof, reseal-able, safety-lock-bags of 100 and 10 bags to a case.