Saw Blades and Frames, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72009 72009-10 72015-01 72015-10 72015-20
102091-240EA 66.19 CAD
102091-240 102097-784 102097-786 102097-788 102097-790
Saw Blades and Frames, Electron Microscopy Sciences
Razors and Blades
Swiss made saw blade used for cutting rubber, plastics, wax, plaster and similar materials which cuts in any direction.
SuperLok™ Diamond Wire starts with tensile core wire
A Swiss made saw frames are lightweight and perfectly balanced. Its ergonomic wooden handle reduces fatigue and provides greater dexterity. Patented blade-lock design eases blade insertion and maintains perfect alignment which eliminates wobble and reduces blade loss during use. It accepts saw blades up to 1.2 mm Ø and 130 mm long. This Frame is used for the Diamond Wire and Spiral Saw Blades.
This wire isMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable