Cocoon Boxes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64300-3 64300-3A 64300-3B 64300-3D 64300-4 64300-4A 64300-4B 64300-6C 64300-6Y 64300-8 64300-8A 64300-9 64300-9A 64300-9B 64302-10 64302-20 64302-30
102095-028EA 35.69 CAD
102095-028 102095-030 102095-032 102095-034 102095-036 102095-038 102095-040 102095-042 102095-044 102095-046 102095-048 102095-050 102095-052 102095-054 102095-058 102095-060 102095-062
Cocoon Boxes, Electron Microscopy Sciences
Boxes Boxes for Vials
The Revolutionary Way for Mailing and Storing Samples
Boxes - Made from polypropylene, envelope shape, available with or without an inner frame. These boxes are the safest way for mailing or storing fragile samples. They are impact and shock resistant With a living-hinge, opaque-clear, reusable and recyclable. The boxes are available in many sizes and styles to fit your needs. Closing Clips and Security Seals are also available as an option.
Vials – are made from polystyrene and they are tough and crystal clear and come with a friction lid. Cocoon Boxes are also available to accept these vials for safe storing and/or mailing valuable samples.
1. BE3, BE3A, BE3B, and BE3D
Box BE3 comes with inner frame to form a cocoon; BE3A is without an inner frame offering more room inside of the box; BE3B comes with 8 round cavities, each 0.94"x0.62"H (24x16mm) for storing Vials DB20; BE3D comes with 6 round cavities each 1.29"x0.62"H (33x16mm) for storing vials DDB30.
2. BE4, BE4A and BE4B
4. BE8 and BE8A
BE8 comes with an inner frame. BE8A - The-Envelope-Mailer - comes without the inner frame.
5.BE9, BE9A, and BE9B
BE9 comes with single inner frame. BE9A comes with two compartments and BE9B comes with 4 compartments.
6. Vials DB10, DB20, and DB30
Vial DB10 is designed for the Cocoon Box BE4B (64300-4B); Vial DB20 is designed for the cocoon Box BE3B (64300-3B); and Vial DB30 is designed for the Cocoon Box BE3D (64300-3D).
7. Locking Clips
Used to secure the cocoon boxes. Clips snap together to keep cocoon boxes securely lock. 100 clips per box.