Photoetched Cover Slips, Round, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72265-12 72265-50
102097-886EA 582.29 CAD
102097-886 102097-888
Photoetched Cover Slips, Round, Electron Microscopy Sciences
Cover Glasses
These coverslips are made from pure white glass and photo etched on the surface for a permanent gridded pattern.
  • Diameter 25 mm
  • Using #2 (0.17 to 0.25 mm) thickness cover slips
  • Unique grid pattern, numbered 1 to 200 squares, each 500 µm width
  • Suitable for insertion into most 6-well plates
  • Adaptable to most microscope mounting plates
  • Facilitates observation of activity growing cells
These coverslips are made from pure white glass and photo etched on the surface for a permanent gridded pattern.