Costar® Brand Cell Culture Clusters, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64705-01 64705-05 64706-01 64706-05 64707-01 64707-05
102095-520EA 702.15 CAD
102095-520 102095-522 102095-524 102095-526 102095-528 102095-530
Costar® Brand Cell Culture Clusters, Electron Microscopy Sciences
Microplates Cell Culture Plates
Tissue culture plates are designed and treated for a wide range of applications. Plates are made from optically clear, medical grade polystyrene with flat bottoms of uniform thickness, allowing for distortion-free observation and microscopic examination. Alphanumeric coordinates are placed on the same focal plane as cell growth-for easy referencing of well position. These plates are also for use with our NetWells and our MeshWell Plates.
  • Maintenance of stock cultures
  • Cell cloning
  • Hybridoma fusions
  • Scale up culture
  • Clone expansion
  • 3H-Thymidine
  • 51Cr Release
More Product Information