CultureWell™ Accessories, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70465-4R 70465-3S 70465-8R 70466-10 70466-30 70465-50R 70465-2R1 70465-2R2 70466-40
100489-174EA 268.36 CAD
100489-174 100489-172 100489-176 100489-178 100489-180 100501-892 100489-168 100489-170 100489-182
CultureWell™ Accessories, Electron Microscopy Sciences
Cover Glasses
Reusable CultureWell™ gaskets from CultureWell™ Chambered Coverglass and CultureWell™ Chamber Coverglass Inserts are available bulk packaged.
  • Gaskets are ideal for forming wells on glass microscope slides or in polystyrene dishes
  • Gaskets are non-sterile and may be sterilized by autoclaving, UV or alcohol