CultureWell™ Chambered Coverglass Plate Inserts, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70461-8R 70461-2R1 70461-4R
100489-164EA 2620.6 CAD
100489-164 100489-156 100489-160
CultureWell™ Chambered Coverglass Plate Inserts, Electron Microscopy Sciences
Cover Glasses
Each CultureWell™ insert is comprised of four chambered coverglass assembled in a disposable frame which is placed in a standard 86 x 128 mm culture plate.
All inserts are sterile and ready to use. The stabilizing insert design and accessory hardware allow fragile coverglass to be easily integrated into protocols for high throughput handling of microscope slides and SBS compliant microtiter plates.
Order Now
SPECIFICATIONS
Environmentally Preferable