CultureWell™ Chambered Coverglass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70460-2R1 70460-2R2 70460-8R 70460-3S 70460-4R 70460-50R
100489-146EA 557.8 CAD
100489-146 100489-148 100501-886 100489-150 100489-152 100489-154
CultureWell™ Chambered Coverglass, Electron Microscopy Sciences
Cover Glasses
Chambered coverglass products are optimal for high resolution microscopy.
  • The thin flat coverglass is the optimal platform for high resolution microscopy and high- sensitivity, quantitative, fluorescent imaging
  • Leak-proof design eliminates well-to-well cross contamination
  • Gaskets stabilize coverglass and provide convenient handling using  conventional microscope slide holders and accessories
  • Gaskets are removable for high resolution imaging
  • Gasket may be re-sterilized and reused
  • Well spacing facilitates liquid handling utilizing multichannel pipettes and robotic dispensers
Chambered coverglass products consists of removable / reusable non-cytotoxic silicone gaskets secured to 24 x 50 mm, #1.5 German coverglass. They may be placed in sterile culture plates or 100 mm diameter culture dishes.