Ultra Diamond Knife, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
30-ULD 35-ULD 40-ULD 15-ULDR 18-ULDR 15-ULDE 21-ULDR 18-ULDE 24-ULDR 21-ULDE 24-ULDE 15-ULD 30-ULDR 30-ULDE 35-ULDR 18-ULD 21-ULD 40-ULDR 35-ULDE 24-ULD 40-ULDE
103306-580EA 13344.19 CAD
103306-580 103306-586 103306-588 103307-082 103307-108 103307-130 103307-134 103307-142 103307-148 103307-152 103307-158 103307-162 103307-166 103307-176 103307-178 103307-182 103307-184 103307-194 103307-196 103307-202 103307-206
Ultra Diamond Knife, Electron Microscopy Sciences
Knives
These knives are available in 35, and 45 degree angles. The optimal thickness range for cutting with these knives is between 30 and 150 nm. They are available in sizes ranging from 1.5 to 4.0 mm as a standard. All of these knives are tested within a thickness range of 30 to 150 nm.
The 45° recognized as the knife for routine sectioning
The 35° diamond knives have been used for reduced compression, smoother section surfaces and better structural preservation. Also these knives have proved advantageous in Lowicryls, inhomogenous samples such as undecalcified bone, dental materials, sectioning of soft industrial samples such as metals and polymers, as well as for hard and brittle samples, for example semiconductors (Si, GaAs, etc), superconducting oxides, nanocrystalline ceramics.
The ultra 35° knife (in the triangular holder) with a cutting range of 100 to 2 µm is used for dry sectioning of epoxy or acrylic resin embedded biological samples, which need to be investigated by element analysis. The gliding of the sections on the dry knife surface is facilitated with the use of our static line II ionizer.
The ultra diamond knife is now available in Jumbo with 35° and 45° angles and in 2.0 and 3.0 mm sizes.